Learn More
Selleck Chemical LLC Cell Counting Kit-8 B34302-5ML
SDP
Supplier:  Selleck Chemical LLC B343025 ML
Encompass_Preferred
The Bimake Cell Counting Kit-8 (CCK-8) is a fluorescent assay for the determination of cell viability in vitro Results are obtained in only 3 steps addition incubation and reading
Catalog No. 50-313-5729