Learn More
Enzo Life Sciences Cell Counting Kit-8 (1x500tests)
SDP
Supplier:  Enzo Life Sciences ALX850039KI01
Encompass_Preferred
Colorimetric kit for simple and accurate cell proliferation and cytotoxicity assays. The Cell Counting Kit-8 is a colorimetric assay kit used to measure cell proliferation and cytotoxicity.It is a ready-to-use solution that does not require radioisotopes and correlates with the [3H]-thymidine incorporation assay. It can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results. CCK-8 has shown to achieve higher sensitivity and stability than MTT, MTS or WST-1. Alternative Name: CCKi-8. Applications: Colorimetric detection. Contents: WST-8 solution, 1-methoxy-PMS
Catalog No. NC9538841