Learn More
BIOLOGIX USA INC CELL CULTURE DISH 60X20MM TRTD
Supplier:  BIOLOGIX USA INC 911615
Encompass_Preferred
Weight: 0.88 lbs▪ Mesh membrane material: Nylon▪Frame material: PS▪ Mesh membrane thickness: 10 ~ 23 μm▪ Membrane pore size: 0.40 μm
Catalog No. NC2445679