Learn More
Research Products International Corp Cell Culture Flask, 25 cm2, Sterile, with Filter Cap, 200 per Case
SDP
Supplier:  Research Products International Corp 162465F
Encompass
Manufactured from ultra clear polystyrene for easy viewing of contents. Proprietary surface treating ensures optimum cell attachment and growth. Additional features include graduated markings on both sides of flask, angled neck design. Supplied sterile, non-pyrogenic, RNase, DNase free and 5 flasks per bag. The flasks are packaged in clear plastic bags.
Hydrophobic filter cap flasks:
Suitable for open culture conditions. Caps contain a hydrophobic membrane with a pore size of 0.22 micrometers, which effectively prevents cross-contamination while meeting the requirements for gas exchange in cell and tissue culture.
Catalog No. 50-136-7606