Learn More
Alkali Scientific ASI™ Cell Culture Flasks, TC Treated, Plugged Cap, 225cm2, 5/Pack, 25/case
SDP
Supplier:  Alkali Scientific TP2225
Encompass_Preferred
ASI™ Cell Culture Flasks provide you with the highest level of protection for your cells during long-term cultivation and cell expansion. The design of the Cell Culture Flasks is dedicated to improving workflow, providing superb protection and bringing peace of mind to scientists when it comes to their research. The Cell Culture Flasks allow you to conduct routine cell culture applications with the peace of mind that comes with knowing that your cell cultures are protected at the highest level. The Cell Culture Flasks are available with cell growth areas ranging from 25 cm ² to 225 cm ² * Angled neck for easy access* Packaged in sterile, zip-sealable bags* High clarity, 100% virgin polystyrene* Frosted writing and clear graduations* Notched bottom for slip free stacking* Sterilized by E Beam* Vented caps (with breathable membrane)* Clear lot number for batch traceability* Non-Pyrogenic, DNase/RNase FREE
Catalog No. 50-302-7140