Learn More
Bellco Glass, Inc. Cell Lifter Narrow Blade
SDP
Supplier:  Bellco Glass, Inc. 229305000
Encompass_Preferred
DescriptionUnique double end design features a 19mm beveled edge flat blade on one end, and Narrow Blade on the otherNarrow Blade is tapered to fit into 96 well plates or micro centrifuge tubes, and can be used to re-suspend stubborn pelletsFlat Blade ensures even contact with growth surface of flasks and dishes Pack DescriptionIndividually Wrapped in BagCase Quantity100SterileYesNon-pyrogenicYesPrimary ColorWhitePrimary MaterialHDPERNase/DNase FreeYes
Catalog No. 50-233-0459