Learn More
Apexbio Technology LLC Cell Mitochondria Isolation Kit I 1kit(50-100 tests)
SDP
Supplier:  Apexbio Technology LLC K27241
Encompass_Preferred
Cell Mitochondria Isolation Kit I is a biochemical kit designed for selective isolation of mitochondria from cultured cells It enables study of mitochondrial physiology and targeted protein profiling by employing differential centrifugation which preserves the structural integrity of isolated mitochondria The kit includes a mitochondrial lysis solution for downstream protein analysis using SDS-PAGE Western blotting or two-dimensional electrophoresis Cytoplasmic proteins depleted of mitochondria collected simultaneously allow investigation into mitochondrial protein release The included Trypan Blue staining solution assists in monitoring cell homogenization effectiveness Based on these features Cell Mitochondria Isolation Kit I holds research potential in mitochondrial bioenergetics metabolic pathway studies and protein trafficking analyses
Catalog No. 50-000-97228