Learn More
STEMCELL Technologies Cell NavigatorTM Golgi Stain
SDP
Supplier:  STEMCELL Technologies 1000837
Encompass
Kit containing C6 NBD ceramide for selectively staining the Golgi apparatus in live or aldehyde-fixed cells - 100 Tests
Catalog No. 50-274-0890