Learn More
Bellco Glass, Inc. Cell Pan& Key Only, 85mL
SDP
Supplier:  Bellco Glass, Inc. 198518500
Encompass_Preferred
Description: Cellector Cell Pan & Key Only 85mL. MADE IN THE USA•Tissue Sieve Homogenizer, used to prepare cell suspensions and gel extractions of proteins•All 304 stainless steel construction•Pan Dimensions: 1-1/4” OD x 2-1/2” W•Complete consists of: Cellector pan & key
Catalog No. 50-194-5225