Learn More
Chemglass Life Sciences Cell Screen, 30 Mesh, SST
SDP
Supplier:  Chemglass Life Sciences CLS5023030
Encompass_Preferred
Cell Screen, 30 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 30 mesh is equivalent to 595 micron and 0.0232 inches.
Catalog No. 50-153-2415