Learn More
Chemglass Life Sciences Cell Screen, 300 Mesh, SST
SDP
Supplier:  Chemglass Life Sciences CLS5023300
Encompass_Preferred
Cell Screen, 300 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only.
Catalog No. 50-153-2423