Learn More
Chemglass Life Sciences Cell Screen, 400 Mesh, SST
SDP
Supplier:  Chemglass Life Sciences CLS5023400
Encompass_Preferred
Cell Screen, 400 Mesh, Stainless Steel . Component of the CLS-5020 Stainless Steel tissue grinder cel dissociation sieves. Pictured are the complete tissue grinders, but this item is cell screen only. 400 mesh is equivalent to 37 micron and 0.0015 inches.
Catalog No. 50-153-2424