Learn More
Pacer Global LLC 70µm Cell Strainer, White, Polypropylene, Individually Wrapped, Sterile, 50 per case
SDP
Supplier:  Pacer Global LLC MLSCTCSS70/DEL
Encompass_Preferred
70μm Cell Strainer, White, Individually Wrapped, Sterile, 50 per case. Polypropylene. Nylon mesh filter. Elongated gripping tab on housing for easy handling. Individually wrapped. Fit most 50ml centrifuge tubes.
Catalog No. NC1946092
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product