Learn More
STEMCELL Technologies CellAdhere Collagen I-Coated
SDP
Supplier:  STEMCELL Technologies 1001199
Encompass
Sterile polystyrene collagen I-coated T-175 cm² flask with vent cap
Catalog No. 50-253-8227