Learn More
Amsbio LLC CELLBANKER 2 (4 x 20 ml)
Supplier:  Amsbio LLC 11893/DEL
Encompass
CELLBANKER 2 is a serum free cell freezing media offering a reliable, consistent and high viability recoveries post-cryopreservation. CELLBANKER 2 contains no animal products.
Catalog No. NC1661791
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.