Learn More
Biotium CellBrite™ NIR790 Cytoplasmic Membrane Dye, 2 mM in DMSO
Supplier:  Biotium 30079
Encompass_Preferred
CellBrite™ NIR Cytoplasmic Membrane Dyes are novel near-infrared carbocyanine dyes for labeling the cytoplasmic membranes of living cells. Due to their long emission wavelengths, near-infrared cell membrane stains can be used to label cells for near-infrared small animal imaging studies for non-invasive imaging of cell migration and cell homing. The dyes have long 18-carbon hydrophobic tails and an additional water-soluble group. These unique chemical structure elements make the dyes easy to dissolve while providing highly stable cytoplasmic membrane staining, unlike traditional carbocyanine dyes like DiI, DiO, and DiR, which are often difficult to dissolve or prone to precipitation during cell staining. CellBrite™ NIR790 has an excitation/emission of 786/820 nm.
Catalog No. 50-196-4500