Learn More
Alkali Scientific CellPro™ 100 mM Sodium Pyruvate solution, 100ml, 10/Case
SDP
Supplier:  Alkali Scientific NAPY01CS
Encompass_Preferred
An easily accessible carbohydrate source for cell culture media. Part of a high-performance line of nutritional and growth supplements
Catalog No. 50-302-8246