Learn More
Alkali Scientific CellPro™ 1L (1000mL) Super-Speed Centrifuge Bottles with Sealing Closure, 6/Case, PPCO
SDP
Supplier:  Alkali Scientific SSC1LPPCO
Encompass_Preferred
Enhance productivity and maximize pellet recovery in high-speed, large-volume separation processes with CellPro™ 1L Super-Speed Centrifuge Bottles. Designed for bacterial, yeast, tissue isolations, and viral harvesting, these bottles deliver exceptional performance in demanding laboratory applications. Process up to 1 liter per bottle at speeds up to 15,810 x g Smooth inner walls minimize shearing in mammalian cell culture separations Polycarbonate option provides superior clarity and mechanical strength Polypropylene option offers excellent chemical resistance Operating temperature range: 4°C to 22°C; storage down to -70°C Three-piece sealing cap assembly (polypropylene plug, closure, and silicone gasket) ensures leakproof† performance USP Class VI compliant, non-cytotoxic, and autoclavable Our CellPro™ bottles match the same high standards and specifications as competing brands, offering a reliable alternative for your laboratory needs.
Catalog No. 50-302-8316