Learn More
Alkali Scientific CellPro™ 1ML Serological Pipets, Indiv. Wrap, 800/cs
SDP
Supplier:  Alkali Scientific SP201CS
Encompass
Serological Pipets • STERILE • Individually Wrapped Serological Pipettes • color coded paper/plastic wrapping for pop-through or peel-apart techniques • Stretched, Non-welded Design • Bi-directional and extra Negative Graduation Marks • Filter Plug and 100% Polystyrene • Gamma Sterilized; Pyrogen/Cytotoxin/RNase & Dnase Free
Catalog No. 50-238-8813