Learn More
Alkali Scientific CellPro™ 96-Well, Silicone Sealing Mats, 50/pk, 96 Square Well, Black Alphanumeric Printing, Silicone, STERILE, "+" Opening, Ø8.4mm, Printed, 50 / PK
SDP
Supplier:  Alkali Scientific SSM33205S
Encompass
CellPro™ 96 Well, Silicone Sealing Mats, 50/pk mat seals 96 wells at a time for minimal evaporation / sample loss, guaranteeing sample integrity, high quality data and uninterrupted workflow. They are useful in PCR, and HTS sample storage and are available in multiple sizes. For a perfect, leakproof seal with most popular plates.
Catalog No. 50-302-8192