Learn More
Alkali Scientific CellPro Culture Test Tube 14mL
SDP
Supplier:  Alkali Scientific CT8236
Encompass_Preferred
Available in Polypropylene or Polystyrene Dual-Position snap caps attached or ordered separately Molded graduations on attached cap versions Sterile RNase DNase pyrogen and cytotoxin free Low density polyethylene (LDPE) snap caps can be attached in two positions The first position allows for the flow of air both in and out of the tube enabling aerobic culture The second position offers a leak-proof and air-tight seal making it ideal for anaerobic culturing and/or sample storage The tubes are available in two plastic polymers Crystal clear polystyrene (PS) tubes withstand temperatures from 0C to 70C while durable transparent polypropylene (PP) tubes withstand temperatures of from -86C to 121C Tubes are available with caps attached and printed graduations or without caps and graduation-free Polystyrene tubes may be centrifuged at up to 2000g Polypropylene tubes withstand higher centrifugal forces up to 4000g All tubes and caps come sterilized to SAL 10-6 and are RNase DNase pyrog
Catalog No. 50-302-9153