Learn More
Alkali Scientific ASI™ Drosophila Bottle Plugs, High-Density Foam, 38.20mm x 31mm, 100/Case
Supplier: Alkali Scientific DF30FB
These high-density foam plugs are specially designed for Drosophila bottles, offering a reliable and efficient solution for laboratories. With a precision die-cut design, they ensure a uniform and snug fit, creating an effective barrier against mites while allowing excellent airflow through the open-cell breathable foam. Perfect for streamlined workflows, these plugs allow gas insertion without removal and eliminate the clinging fibers commonly associated with cotton plugs. Reusable, washable, and fully autoclavable, they are a cost-effective choice for labs seeking durability and performance. Their strong and flexible construction ensures they remain secure during use, even in demanding environments. Packaged in a case of 100, these bottle plugs are ideal for labs of any size that value convenience, reliability, and affordability. Simplify your lab processes and enhance efficiency with these premium high-density foam plugs.