Learn More
Alkali Scientific CellPro™ LB + Ampicillin 100µg/mL + Chloramphenicol 50µg/mL + Tetracycline 25µg/mL, 10/Sleeve
SDP
Supplier:  Alkali Scientific DC0063
Encompass
CellPro™ LB + Ampicillin is a specialized bacterial growth medium designed for the cultivation of ampicillin-resistant bacterial strains. This nutrient-rich agar contains ampicillin, an antibiotic that inhibits the growth of bacteria lacking ampicillin resistance genes. Ideal for molecular biology and microbiology applications, this medium promotes the selective growth of genetically modified bacteria harboring ampicillin resistance plasmids, facilitating the isolation and maintenance of recombinant DNA constructs.
Catalog No. 50-238-8792