Learn More
Alkali Scientific CellPro™ LB, Miller, Chloramphenicol 12.5µg/mL, 10/Sleeve, 150mm
SDP
Supplier:  Alkali Scientific DC0155
Encompass
CellPro™ LB Agar, Miller is a solid medium commonly used for the cultivation and maintenance of Escherichia coli (E. coli) and other recombinant bacterial strains. This agar variant contains tryptone, yeast extract, sodium chloride, and agar, providing essential nutrients for bacterial growth. LB Agar, Miller is particularly suitable for molecular biology and microbiology applications, including the selection and propagation of transformed bacterial cells carrying recombinant DNA constructs.
Catalog No. 50-238-9443