Learn More
Alkali Scientific CellPro Metal Free Centrifuge
SDP
Supplier:  Alkali Scientific CTMF2915
Encompass
Total-PureCentrifuge Tubes are produced from a proprietary purified polypropylene blend that is certified to be free of trace metal content Tested via ICP-MS these tubes are certified to contain less than 1ppm of common trace metals including Calcium Magnesium Zinc Iron Manganese Copper Aluminum Silicon Nickel Vanadium Sodium Phosphorous Cobalt Chromium Potassium Lithium Lead Selenium Cadmium Mercury and Arsenic In addition to a superior level of purity TotalPureTubes include a wide array of premium features These tubes are engineered to withstand a max centrifugation rate of 20000xg with this RCF specification printed directly on both the 15mL and 50mL tubes These tubes also feature Hi-Vizgraduations that are easily identified at any angle and the printed max-freeze line indicates the maximum allowable volume to be frozen inside the tubes TotalPures natural color caps offer a smooth seal that is easy to open/close with one hand without compromising the integrity of the 95kPA leak
Catalog No. 50-302-7302