Learn More
Alkali Scientific CellPro Premium Cell Strainer
SDP
Supplier:  Alkali Scientific MT6050
Encompass
The SureStrainPremium Cell Strainer system is a fast and easy-to-use solution for detaching isolating and straining cells from clusters and tissues in preparation for flow cytometry A polypropylene frame supports a nylon mesh that is available in eight color-coded pore sizes ranging from 5 m to 100 m The 5 new pore sizes allow for finer control over cell aggregate and clump filtration The advanced polypropylene frame design also contributes to superior performance and reduced processing time by eliminating the air-lock effect Improved aeration allows air to escape while filtrate passes through the nylon mesh into the tube preventing messy spillovers 8 pore sizes for greater cell sorting precision Advanced design faster flow through Included adapter for tubes smaller than 50mL Compatible with 6-well plates Interlocking and stackable 1 5mm feet for straining into flat plates and dishes Sterile individual blister packaging RNase DNase pyrogen and cytotoxin free
Catalog No. 50-302-6795