Learn More
Alkali Scientific CellPro™ Screw Cap Culture Tubes with Phenolic Caps and White Marking Area, Flat Bottom, Silicone Rubber Liner, 11mL, 16x100mm, 100/Pk
SDP
Supplier:  Alkali Scientific STSC11F
Encompass_Preferred
CellPro™ Screw Cap Culture Tubes provide dependable performance for sample handling, storage, and culturing in microbiology, clinical, and laboratory research environments. Constructed from high quality glass, these tubes are equipped with durable phenolic screw caps that create a secure, leak resistant seal and withstand repeated sterilization and routine laboratory handling. The flat bottom design allows the tubes to stand upright on benches and work surfaces without the need for additional racks, improving stability during filling, labeling, and storage. A clearly defined white marking area allows accurate sample identification and traceability for organized laboratory workflows. Compatible with standard racks and accessories, these culture tubes are ideal for routine culturing, reagent storage, and sample preparation. Each tube includes a silicone rubber liner that improves sealing reliability and chemical resistance.
Catalog No. 50-006-62918