Learn More
Alkali Scientific CellPro™ SOC Media 10 x 10mL
SDP
Supplier:  Alkali Scientific DC0002
Encompass_Preferred
Alkali Scientific™ CellPro™ SOC Media is a specialized growth medium designed to support the transformation and propagation of Escherichia coli cells. Formulated with essential nutrients and salts, this media promotes rapid cell growth and high transformation efficiency, making it ideal for molecular cloning and recombinant DNA techniques. Through meticulous sterile filtration, CellPro™ SOC Media maintains purity and consistency, ensuring optimal performance in molecular biology laboratories.
Catalog No. 50-238-8670