Learn More
Alkali Scientific CellPro™ TAE, 50X (Tris Acetate EDTA), 1L, 12/cs
SDP
Supplier:  Alkali Scientific DB0332CS
Encompass
Alkali Scientific™ CellPro™ TAE Tris Acetate EDTA is a fundamental reagent in nucleic acid electrophoresis, renowned for its exceptional buffering capacity and DNA solubilization properties. Designed to provide optimal performance and reproducibility, this solution enables precise separation and analysis of DNA fragments in agarose gel electrophoresis. Each batch undergoes rigorous sterile filtration to ensure purity and consistency, empowering researchers with confidence in their molecular biology experiments.
Catalog No. 50-238-8923