Learn More
Alkali Scientific CellPro™ Tip Comb, 96 Well, V-Bottom, PP, 50 bags of 2 pieces, 100 / CS, 96, V-Bottom, PP, STERILE, for Kingfisher™ and IsoPURE™, 50 bags of 2 pieces, 100 / CS
SDP
Supplier:  Alkali Scientific TC334096S
Encompass_Preferred
CellPro™ 96 Well, V-Bottom Tip Combs are made specifically to fit KingFisher™ Flex systems and for a variety of other automation platforms. Produced from low-binding, medical grade polypropylene, they are available sterile or nonsterile. Applications include DNA extraction, quantification, viral RNA extraction, immunoprecipitations, etc.  • Compatible with KingFisher™ Flex• Low-binding, medical-grade polypropyleneZ• RNase, DNase free
Catalog No. 50-302-8933