Learn More
Alkali Scientific CellPro™ Tryptic Soy Broth, 90mL, 48/Case
SDP
Supplier:  Alkali Scientific TSB0190
Encompass_Preferred
CellPro™ Tryptic Soy Broth (TSB), 90mL, is a nutrient rich, general purpose liquid culture medium formulated to support the growth and recovery of a broad range of aerobic and facultative anaerobic microorganisms. Prepared with enzymatic digests of casein and soybean meal, this versatile medium provides essential nutrients that promote reliable microbial proliferation. It is widely used for sterility testing, environmental monitoring, microbial enrichment, culture maintenance, and quality control applications in clinical, pharmaceutical, food, and industrial laboratories. The ready to use 90mL format simplifies laboratory workflows by reducing preparation time and minimizing variability between batches. Manufactured under strict quality standards, CellPro™ TSB delivers consistent performance and dependable results for routine microbiological testing and research applications.
Catalog No. 50-006-62453