Learn More
Alkali Scientific CellPro™ Trypticase Soy Agar (TSA) Plates 100mm, 10/Sleeve
SDP
Supplier:  Alkali Scientific TSAP01
Encompass
CellPro™ Trypticase Soy Agar (TSA) Plates, 100mm, are general purpose microbiological culture media designed for the cultivation and isolation of a wide variety of aerobic and facultative anaerobic microorganisms. Formulated with enzymatic digests of casein and soybean meal, TSA provides a nutrient rich environment that supports the growth of both fastidious and non fastidious organisms. These ready to use plates are widely used in microbiology laboratories for environmental monitoring, sterility testing, microbial enumeration, and routine culture maintenance. The 100mm plate format offers ample surface area for colony development and easy observation. Manufactured under strict quality standards, CellPro™ TSA plates deliver reliable performance and consistent results for clinical, pharmaceutical, food safety, and research laboratory applications.
Catalog No. 50-006-62779