Learn More
Alkali Scientific CellPro™ Tryptone + Top Agar 0.7% 100mL
SDP
Supplier:  Alkali Scientific DC0012
Encompass_Preferred
Alkali Scientific™ CellPro™ Tryptone is a high-quality peptone derived from casein, widely used as a nutrient source in microbial culture media. With its rich amino acid composition, this molecular biology-grade tryptone supports robust bacterial growth and protein expression. Each batch undergoes stringent sterile filtration to ensure purity and consistency, making CellPro™ Tryptone an essential component in various laboratory applications, including bacterial culture, recombinant protein production, and microbiological research.
Catalog No. 50-238-8740