Learn More
Partec North America CELLTRICS 20UM STERILE SING PK
Supplier:  Partec North America 040042325
Encompass
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures.
Catalog No. NC9699018