Learn More
Partec North America Sterile, single-pack CellTrics™ filters, 30um (50/box)
Supplier: Partec North America 040042326
CellTrics can be used to filter any kind of liquid suspension, e.g. for isolation of cells and nuclei from cell debris and aggregates and also for filtering either microorganisms or cell cultures. 50/PK.