Learn More
Alkali Scientific Cellulase [Onozuka R 10], 5 G
SDP
Supplier:  Alkali Scientific CEL003CP
Encompass_Preferred
Cellulase Onozuka R10, 5 g, is an enzyme grade reagent designed to catalyze the breakdown of cellulose in plant cell walls. It is commonly used in a variety of research applications, including plant cell culture, genetic engineering, and the extraction of intracellular components. By degrading cellulose, this cellulase enzyme facilitates the release of valuable compounds, making it ideal for experiments in plant biology, biochemistry, and agriculture. It is also employed in industrial applications, such as biofuel production and the processing of plant materials. This high-purity cellulase enzyme is a crucial tool in both laboratory research and commercial applications that require efficient cell wall digestion.
Catalog No. 50-006-60379