Learn More
AG Scientific Inc Cellulase [Onozuka R-10] 5 G
Supplier:  AG Scientific Inc C27455GM
Encompass_Preferred
Cellulase [Onozuka R-10] is derived from Trichoderma Viride. Cellulase R-10 is a multi-enzymatic system consisting of various enzymes, including cellulase, hemicellulase, pectinase and protease.Cellulase R-10 is typically used in association with macerozyme R-10 in the decomposition of plant cell walls to produce protoplasts.
Catalog No. 50-226-8273