Learn More
Medchemexpress LLC Cellulose, 2-hydroxypropyl ether | 9004-64-2 | 97.0% | 500 G
SDP
Supplier:  Medchemexpress LLC HY100557500G
Encompass_Preferred
Hydroxypropyl cellulose is a non-ionic cellulose ether supplied as a fine powder for use as a pharmaceutical excipient and laboratory reagent. It functions as a thickener and viscosifier and is commonly used in coatings, emulsions, suspensions, and tablet formulations. Certificate of analysis and safety data sheet are available from the supplier.
Catalog No. 50-002-11604