Learn More
Tisch Scientific Tisch Scientific Membrane Filter, Cellulose Acetate (CA), 0.45 µm, 47 mm, Nonsterile, 200 Pack, SF15103
SDP
Supplier:  Tisch Scientific SF15103
Encompass
Tisch cellulose acetate membrane filters are a hydrophilic filtration option perfect for use in aqueous protein solutions and ground water filtration applications.
Catalog No. NC1776561