Learn More
Medchemexpress LLC Cenicriviroc | 497223-25-3 | 98.1% | 696.94 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1488210MM/1ML
Encompass_Preferred
Cenicriviroc is an orally active compound that functions as a dual antagonist of CCR2 and CCR5. Its biological activity includes the inhibition of both HIV-1 and HIV-2. Furthermore, it exhibits potent anti-inflammatory and antiinfective properties, indicating its potential application in treating conditions related to these activities.
  • Orally active.
  • Dual antagonist of CCR2/CCR5.
  • Inhibits HIV-1 and HIV-2.
  • Potent anti-inflammatory activity.
  • Potent antiinfective activity.
Catalog No. 50-003-60847