Learn More
Medchemexpress LLC Cenicriviroc Mesylate | 497223-28-6 | 99.9% | 793.05 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14882A
Encompass_Preferred
Cenicriviroc Mesylate is a dual CCR2/CCR5 antagonist that inhibits both HIV-1 and HIV-2. It exhibits potent anti-inflammatory and antiinfective activity, making it suitable for various research applications.
  • Dual CCR2/CCR5 antagonist
  • Inhibits HIV-1 and HIV-2
  • Potent anti-inflammatory activity
  • Potent antiinfective activity
  • Active in cellular models (IC50 for CCR5: 0.29 nM, CCR2: 5.9 nM)
  • Reduces monocyte/macrophage recruitment in vivo
Catalog No. 50-003-63526