Learn More
Polycarbin 50 mL Centrifuge Tube CS500, Racked (Sterile)
SDP
Supplier:  Polycarbin PC010101G
Encompass_Preferred
Polycarbin's racked 50 mL conical tubes are made using 20% circular economy, post-consumer recycled content. Each lot of tubes is sterile. Certified DNase, RNase, and Pyrogen free. 25 tubes/rack, 20 racks/case, 500 tubes/case. Centrifugation Speed 12,000 x g. Temperature Range of -196°C to 100°C. Manufactured in a ISO 9001 compliant facility.
Catalog No. NC2213713