Learn More
Cayman Chemical Centaurea maculosa CnIcIn 5mg
SDP
Supplier:  Cayman Chemical 389695 MG
Encompass_Preferred
A germacranolide sesquiterpene lactone with diverse biological activities; irreversibly inhibits MurA (IC50s = 16.7 and 10.3 for the E. coli and P. aeruginosa enzymes, respectively) and is active against B. cereus and M. flavus (MICs = 1.25 and 5 UG/ml, respectively); inhibits the growth of MCF-7 and OVCAR-3 cancer cells (GI50s = 37.8 and 37.4 µM, respectively); reduces snake venom-induced paw edema in rats from 2.5-10 mg/kg
Catalog No. 50-291-6251