Learn More
Grainger Center Drill Plain 1.10mm
Supplier:  Grainger 16T474
Encompass_Preferred
The Grainger center drills are made of high speed steel and have a 60 degree included angle point. They are available in multiple fractional drill sizes from 1/16" up to 1/2". The drills have a straight shank and come uncoated. Overall length ranges from 2" for the smaller sizes up to 5" for the larger sizes. These center drills are designed for starting holes in preparation for drilling and produce a conical shaped hole.
Catalog No. 50-276-8275