Learn More
Grainger Center Drill Plain 1.30mm
Supplier:  Grainger 16T475
Encompass_Preferred
The FisherbrandTM 502766030 is a plastic, disposable, sterile, serological pipette with a capacity of 10 mL. It has graduations of 0.1 mL and a length of 230 mm. The pipette is individually wrapped and gamma irradiated. It can be used for routine serological techniques. The pipettes are free of detectable DNase, RNase, DNA and ATP. They are non-pyrogenic and non-cytotoxic. The pipettes are made in ISO 9001 certified facilities.
Catalog No. 50-276-6030