Learn More
Thomas Scientific Centricon Plus 70 100K NMWL 8pk
SDP
Supplier:  Thomas Scientific 1211L15
Encompass
This centrifugal filter enables the concentration of proteins with a membrane NMWL of 100 kDa. Centricon centrifugal filters are ideal for concentrating & desalting proteins in sample volumes from 15 to 70mL. Centricon Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions Device consists of a polypropylene filtrate collection cup, sample filter cup, concentrate/retentate cup and cap Concentrate 70 mL to 350 µL in just 25 minutes
Catalog No. NC1092068