Learn More
Medchemexpress LLC Cep dipeptide 1 | 816432-15-2 | 98.5% | 381.42 | 100 MG

Supplier: Medchemexpress LLC HY16959100MG
CEP dipeptide 1 is a CEP dipeptide that exhibits potent angiogenic activity. It acts as a mediator of age-related macular degeneration (AMD). This product appears as an off-white to yellow solid and is recommended for research use only, not intended for patient use.
- Solubility in H2O: 10 mg/mL (26.22 mM); requires sonication.
- Store sealed and away from moisture.
- Powder storage: -80°C for 2 years, -20°C for 1 year.
- In solvent storage: -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.