Learn More
Cayman Chemical CEP-LysIn-d4 1mg
SDP
Supplier:  Cayman Chemical 90005951 MG
Encompass_Preferred
An internal standard for the quantification of CEP-Lysine by GC- or LC-MS
Catalog No. 50-289-4531