Learn More
Cayman Chemical CephacetrIle 5mg
SDP
Supplier:  Cayman Chemical 362695 MG
Encompass_Preferred
A cephalosporin antibiotic; active against clinical isolates of S. aureus, S. epidermidis, S. pyogenes, E. coli, and Klebsiella (MICs = 0.64, 0.64, 0.16, 6.25 and 3.12 UG/ml, respectively); active against veterinary isolates of S. aureus from bovine mastitis at 30 UG/disc
Catalog No. 50-291-2589