Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cephaeline dihydrochloride | 5853-29-2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYN226050MG

Encompass_Preferred

Cephaeline dihydrochloride is a selective CYP2D6 inhibitor with an IC50 of 121 μM. It is for research use only and is not sold to patients. This compound demonstrates significant metabolic activity for the generation of 9-O-demethylEmetine and Cephaeline.

  • Selective CYP2D6 inhibitor
  • IC50 of 121 μM against CYP2D6
  • Inhibits CYP2C9 and CYP3A4 with IC50s over 1000 μM
  • Ki for CYP2D6 activity is 54 μM
  • Ki for CYP3A4 activity is 355 μM
  • Appearance: white to off-white solid
  • Purity: 98.95%

Catalog No. 50-004-17794


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.